Skip to Content
Merck
All Photos(4)

Key Documents

SAB2100658

Sigma-Aldrich

Anti-EIF2AK1 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-Eukaryotic translation initiation factor 2-α kinase 1, Anti-HRI, Anti-KIAA1369

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
R$3,345.00

R$3,345.00


Check Cart for Availability


Select a Size

Change View
100 μL
R$3,345.00

About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

R$3,345.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

71 kDa

species reactivity

dog, guinea pig, horse, human, mouse, rabbit, rat, bovine

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... EIF2AK1(27102)

Immunogen

Synthetic peptide directed towards the N terminal region of human EIF2AK1

Biochem/physiol Actions

EIF2AK1 acts at the level of translation initiation to downregulate protein synthesis in response to stress. The protein is a kinase that can be inactivated by hemin. Two transcript variants encoding different isoforms have been found for this gene.The HRI gene is localized to 7p22 where its 3′ end slightly overlaps the 3′ end of the gene JTV1. The two genes are transcribed from opposite strands. Studies in rat and rabbit suggest that the HRI gene product phosphorylates the alpha subunit of eukaryotic initiation factor 2. Its kinase activity is induced by low levels of heme and inhibited by the presence of heme. Sequence Note: The sequence AF181071.1 is a chimeric mRNA clone. Only the heme-regulated initiation factor 2-alpha kinase region was propagated into this RefSeq record.

Sequence

Synthetic peptide located within the following region: TCSDEFSSLRLHHNRAITHLMRSAKERVRQDPCEDISRIQKIRSREVALE

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service