Skip to Content
Merck
All Photos(1)

Key Documents

SAB2100371

Sigma-Aldrich

Anti-CCRL2 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-CKRX, Anti-CRAM-A, Anti-CRAM-B, Anti-Chemokine (C-C motif) receptor-like 2, Anti-HCR

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
R$3,345.00

R$3,345.00


Check Cart for Availability


Select a Size

Change View
100 μL
R$3,345.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

R$3,345.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

39 kDa

species reactivity

mouse, human

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CCRL2(9034)

Immunogen

Synthetic peptide directed towards the N terminal region of human CCRL2

Biochem/physiol Actions

CCRL2 is a chemokine receptor like protein, which is predicted to be a seven transmembrane protein and most closely related to CCR1. Chemokines and their receptors mediated signal transduction are critical for the recruitment of effector immune cells to the site of inflammation. CCRL2 gene is expressed at high levels in primary neutrophils and primary monocytes, and is further upregulated on neutrophil activation and during monocyte to macrophage differentiation. The function of this gene is unknown. This gene is mapped to the region where the chemokine receptor gene cluster is located.This gene encodes a chemokine receptor like protein, which is predicted to be a seven transmembrane protein and most closely related to CCR1. Chemokines and their receptors mediated signal transduction are critical for the recruitment of effector immune cells to the site of inflammation. This gene is expressed at high levels in primary neutrophils and primary monocytes, and is further upregulated on neutrophil activation and during monocyte to macrophage differentiation. The function of this gene is unknown. This gene is mapped to the region where the chemokine receptor gene cluster is located.

Sequence

Synthetic peptide located within the following region: LSAQLVPSLCSAVFVIGVLDNLLVVLILVKYKGLKRVENIYLLNLAVSNL

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mélodie Diot et al.
Reproductive biology and endocrinology : RB&E, 13, 81-81 (2015-08-01)
In mammals, adipose tissue is able to secrete various hormones called adipokines including adiponectin (ADP), chemerin (Chem) and visfatin (Visf) which are involved in controlling energy metabolism as well as reproductive functions. Visf receptor is still unknown whereas ADP and
Anthony Estienne et al.
Poultry science, 102(1), 102248-102248 (2022-11-25)
Chemerin is a hormone produced mainly by adipose tissue and liver. We have recently shown that it is locally produced in the reproductive tract in hens, particularly at the magnum level, leading to its accumulation in the egg albumen. We
Edyta Rytelewska et al.
Molecular reproduction and development (2020-06-20)
Recent studies have demonstrated that chemerin participates in the regulation of female reproductive function at the level of the ovaries. Due to the lack of data concerning the presence of the chemerin system (chemerin and its receptors: CMKLR1, GPR1, CCRL2)
Malgorzata Brzoskwinia et al.
International journal of molecular sciences, 21(12) (2020-06-26)
Adipokines influence energy metabolism and have effects on male reproduction, including spermatogenesis and/or Sertoli cell maturation; however, the relationship between these active proteins and androgens in testicular cells is limited. Here, we studied the impact of short-term exposure to flutamide

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service