Skip to Content
Merck
All Photos(1)

Key Documents

SAB1412937

Sigma-Aldrich

Anti-GAS2L2 antibody produced in mouse

purified immunoglobulin

Synonym(s):

Anti-GAR17

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

form

buffered aqueous solution

mol wt

antigen 96.8 kDa

species reactivity

human

technique(s)

western blot: 1 μg/mL

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GAS2L2(246176)

Immunogen

GAS2L2 (NP_644814.1, 1 a.a. ~ 880 a.a) full-length human protein.

Sequence
MSQPAGGRRKPRTLGPPVCSIRPFKSSEQYLEAMKEDLAEWLRDLYGLDIDAANFLQVLETGLVLCQHANVVTDAALAFLAEAPAQAQKIPMPRVGVSCNGAAQPGTFQARDNVSNFIQWCRKEMGIQEVLMFETEDLVLRKNVKNVVLCLLELGRRAWRFGVAAPTLVQLEEEIEEEVRRELALPPPDPSPPAPPRRQPCHFRNLDQMVQSLVSHCTCPVQFSMVKVSEGKYRVGDSNTLIFIRILRNHVMVRVGGGWDTLGHYLDKHDPCRCTSLSHKPGSFLKPPAPPVQHEVRVQDGPSQTQPTMTISRSQSPPPPVDWKTYTSSDRRLRPPTPSSPRPRRERGAGTGASREMAPFLRCQERSLIPSWRQPTAGDSPPSPQSSSTQKGRDPQCTSSGKREERYPPELPRGRIPTSWVHEETDSWGTDAGNPTPQRLRAIEATTKGISARGPSPLPRSFGPAECLGLRLPLRDEAKGAFFQFREPESVRSPTPVQGLTKIPIRLPPARPPTPGRSFPGATSGSPRTELGRDPIPLRAVTVDLAGSTHGDCSVEVRQEDQQLDIQVMAEARESWDLGLQEQEGRYTPLPLGGNKEQAIYCSLEEEILGNMKLLEVRSACPQGTRSGVIPRSGVYIPRLAGQWPEPGGPYDKAIQELAQGSPSLLKVDLEAWKAAPTGSPKPAVTPGPGSLKGKLGARQSGPRTKASLSAKGTHMRKVPPQGGQDCSASTVSASPEAPTPSPLDPNSDKAKACLSKGRRTLRKPKRVPSIYKLKLRPRIRPRRDHRPEKQPSRIPRPLAYVFLGPARQPPKDRLLRAVLGSKGGEASRVDGASVGEEEEEGKEEKEPAAPLESSPQPPEGLQPHWLNQAPLPPEEESWV

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service