Skip to Content
Merck
All Photos(2)

Key Documents

SAB1412773

Sigma-Aldrich

Monoclonal Anti-SYT7 antibody produced in mouse

clone 4H4, purified immunoglobulin

Synonym(s):

Anti-IPCA-7, Anti-MGC150517, Anti-PCANAP7, Anti-SYT-VII

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
R$2,669.00

R$2,669.00


Check Cart for Availability


Select a Size

Change View
100 μG
R$2,669.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

R$2,669.00


Check Cart for Availability

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

4H4, monoclonal

form

buffered aqueous solution

mol wt

antigen 36.63 kDa

species reactivity

human

technique(s)

ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG2a

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SYT7(9066)

General description

Synaptotagmins, such as SYT7, are brain-specific calcium-dependent phospholipid-binding proteins that play a role in synaptic exocytosis and neurotransmitter release. See MIM 600782.[supplied by OMIM

Immunogen

SYT7 (NP_004191.2, 41 a.a. ~ 139 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
CQRKLGKRYKNSLETVGTPDSGRGRSEKKAIKLPAGGKAVNTAPVPGQTPHDESDRRTEPRSSVSDLVNSLTSEMLMLSPGSEEDEAHEGCSRENLGRI

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bing Xiao et al.
Molecular medicine reports, 16(6), 9017-9022 (2017-10-11)
Synaptotagmin‑7 (SYT7) is a member of the synaptotagmin gene family, and encodes a protein that mediates the calcium‑dependent regulation of membrane trafficking during synaptic transmission. A previous study demonstrated that the expression of SYT7 is associated with prostate cancer and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service