SAB1412722
Monoclonal Anti-KCNJ5 antibody produced in mouse
clone 8D2, purified immunoglobulin
Synonym(s):
Anti-CIR, Anti-GIRK4, Anti-KATP1, Anti-KIR3.4
About This Item
biological source
mouse
conjugate
unconjugated
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
8D2, monoclonal
form
buffered aqueous solution
mol wt
antigen 36.63 kDa
species reactivity
human
technique(s)
ELISA: suitable
western blot: 1-5 μg/mL
isotype
IgG2a
GenBank accession no.
UniProt accession no.
shipped in
dry ice
storage temp.
−20°C
target post-translational modification
unmodified
Gene Information
human ... KCNJ5(3762)
General description
Immunogen
Sequence
SYMDTEVLWGHRFTPVLTLEKGFYEVDYNTFHDTYETNTPSCCAKELAEMKREGRLLQYLPSPPLLGGCAEAGLDAEAEQNEEDEPKGLGGSREARGSV
Features and Benefits
Physical form
Legal Information
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service