Skip to Content
Merck
All Photos(1)

Key Documents

SAB1411597

Sigma-Aldrich

Anti-TAF1C antibody produced in rabbit

purified immunoglobulin, buffered aqueous solution

Synonym(s):

MGC:39976, SL1, TAFI110, TAFI95

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
R$3,595.00

R$3,595.00


Check Cart for Availability


Select a Size

Change View
100 μG
R$3,595.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

R$3,595.00


Check Cart for Availability

biological source

rabbit

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 85.1 kDa

species reactivity

human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TAF1C(9013)

General description

Initiation of transcription by RNA polymerase I requires the formation of a complex composed of the TATA-binding protein (TBP) and three TBP-associated factors (TAFs) specific for RNA polymerase I. This complex, known as SL1, binds to the core promoter of ribosomal RNA genes to position the polymerase properly and acts as a channel for regulatory signals. This gene encodes the largest SL1-specific TAF. Two transcripts encoding different isoforms have been identified. (provided by RefSeq)

Immunogen

TAF1C (AAH28131.1, 1 a.a. ~ 775 a.a) full-length human protein.

Sequence
MLPPLIDPWDPGLTARDLLFRGGYRYRKRPRVVLDVTEQISRFLLDHGDVAFAPLGKLMLENFKLEGAGSRTKKKTVVSVKKLLQDLGGHQPWGCPWAYLSNRQRRFSILGGPILGTSVASHLAELLHEELVLRWEQLLLDEACTGGALAWVPGRTPQFGQLVYPAGGAQDRLHFQEVVLTPGDNPQFLGKPGRIQLQGPVRQVVTCTVQGETLLAVRSDYHCAVWKFGKQWQPTLLQAMQVEKGATGISLSPHLPGELAICSRSGAVCLWSPEDGLRQIYRDPETLVFRDSSSWRWADFTAHPRVLTVGDRTGVKMLDTQGPPGCGLLLFRLGAEASCQKGERVLLTQYLGHSSPKCLPPTLHLVCTQFSLYLVDERLPLVPMLKWNHGLPSPLLLARLLPPPRPSCVQPLLLGGQGGQLQLLHLAEGASVPRLAGPPQSLPSRIDSLPAFPLLEPKIQWRLQERLKAPTIGLAAVVPPMPSAPTPGLVLFQLSAAGDVFYQQLRPQVDSSLRRDAGPPGDTQPDCHAPTASWTSQDTAGCSQWLKALLKVPLAPPVWTAPTFTHRQMLGSTELRREEEEGQRLGVLRKAMARGQLLLQRDLGSLPAAEPPPAPESGLEDKLSERLGEAWAGRGAAWWERQQGRTSEPGRQTRRPKRRTQLSSSFSLSGHVDPSEDTSSPHSPEWPPADALPLPPTTPPSQELTPDACAQGVPSEQRQMLRDYMAKLPPQRDTPGCATTPPHSQASSVRATRSQQHTPVLSSSQPLRKKPRMGF

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service