Skip to Content
Merck
All Photos(1)

Key Documents

SAB1410671

Sigma-Aldrich

Anti-APOA2 antibody produced in rabbit

purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-Apo-AII, Anti-ApoA-II, Anti-Apolipoprotein A-II, Anti-Apolipoprotein A2, Anti-apoA-II

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
R$3,595.00

R$3,595.00


Check Cart for Availability


Select a Size

Change View
100 μG
R$3,595.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

R$3,595.00


Check Cart for Availability

biological source

rabbit

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 11.2 kDa

species reactivity

human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... APOA2(336)

General description

This gene encodes apolipoprotein (apo-) A-II, which is the second most abundant protein of the high density lipoprotein particles. The protein is found in plasma as a monomer, homodimer, or heterodimer with apolipoprotein D. Defects in this gene may result in apolipoprotein A-II deficiency or hypercholesterolemia. (provided by RefSeq)

Immunogen

APOA2 (NP_001634.1, 1 a.a. ~ 100 a.a) full-length human protein.

Sequence
MKLLAATVLLLTICSLEGALVRRQAKEPCVESLVSQYFQTVTDYGKDLMEKVKSPELQAEAKSYFEKSKEQLTPLIKKAGTELVNFLSYFVELGTQPATQ

Physical form

Solution in phosphate buffered saline, pH 7.4

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. Anti-ApoA2antibodyの購入を考えております。immunoglobulinにしてはSDS-PAGEにおいて分子量が小さすぎますが、なぜでしょうか? 抗原側の販売ということでしょうか?

    1 answer
    1. The immunogen used to develop this antibody is the APOA2 (NP_001634.1, 1 a.a. ~ 100 a.a) full-length human protein with a molecular weight of 11.2 kDa. The molecular weight indicated is not for the antibody, it is for the protein. See the enlarged image below for details:

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service