Skip to Content
Merck
All Photos(4)

Documents

SAB1410064

Sigma-Aldrich

Anti-TRAP1 antibody produced in rabbit

purified immunoglobulin, buffered aqueous solution

Synonym(s):

HSP75, HSP90L

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 80 kDa

species reactivity

mouse, human

technique(s)

immunofluorescence: suitable
western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TRAP1(10131)

General description

HSP90 proteins are highly conserved molecular chaperones that have key roles in signal transduction, protein folding, protein degradation, and morphologic evolution. HSP90 proteins normally associate with other cochaperones and play important roles in folding newly synthesized proteins or stabilizing and refolding denatured proteins after stress. TRAP1 is a mitochondrial HSP90 protein. Other HSP90 proteins are found in cytosol (see HSP90AA1; MIM 140571) and endoplasmic reticulum (HSP90B1; MIM 191175) (Chen et al., 2005 [PubMed 16269234]).[supplied by OMIM

Immunogen

TRAP1 (NP_057376.1, 1 a.a. ~ 704 a.a) full-length human protein.

Sequence
MARELRALLLWGRRLRPLLRAPALAAVPGGKPILCPRRTTAQLGPRRNPAWSLQAGRLFSTQTAEDKEEPLHSIISSTESVQGSTSKHEFQAETKKLLDIVARSLYSEKEVFIRELISNASDALEKLRHKLVSDGQALPEMEIHLQTNAEKGTITIQDTGIGMTQEELVSNLGTIARSGSKAFLDALQNQAEASSKIIGQFGVGFYSAFMVADRVEVYSRSAAPGSLGYQWLSDGSGVFEIAEASGVRTGTKIIIHLKSDCKEFSSEARVRDVVTKYSNFVSFPLYLNGRRMNTLQAIWMMDPKDVGEWQHEEFYRYVAQAHDKPRYTLHYKTDAPLNIRSIFYVPDMKPSMFDVSRELGSSVALYSRKVLIQTKATDILPKWLRFIRGVVDSEDIPLNLSRELLQESALIRKLRDVLQQRLIKFFIDQSKKDAEKYAKFFEDYGLFMREGIVTATEQEVKEDIAKLLRYESSALPSGQLTSLSEYASRMRAGTRNIYYLCAPNRHLAEHSPYYEAMKKKDTEVLFCFEQFDELTLLHLREFDKKKLISVETDIVVDHYKEEKFEDRSPAAECLSEKETEELMAWMRNVLGSRVTNVKVTLRLDTHPAMVTVLEMGAARHFLRMQQLAKTQEERAQLLQPTLEINPRHALIKKLNQLRASEPGLAQLLVDQIYENAMIAAGLVDDPRAMVGRLNELLVKALERH

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service