Skip to Content
Merck
All Photos(5)

Key Documents

SAB1408720

Sigma-Aldrich

Monoclonal Anti-SGK2 antibody produced in mouse

clone 4G4, purified immunoglobulin, buffered aqueous solution

Synonym(s):

H-SGK2, dJ138B7.2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
R$3,595.00

R$3,595.00


Estimated to ship onApril 25, 2025



Select a Size

Change View
100 μG
R$3,595.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

R$3,595.00


Estimated to ship onApril 25, 2025


biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

4G4, monoclonal

form

buffered aqueous solution

mol wt

antigen 33.88 kDa

species reactivity

human, mouse

technique(s)

indirect ELISA: suitable
indirect immunofluorescence: suitable
western blot: 1-5 μg/mL

isotype

IgG3κ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SGK2(10110)

General description

This gene encodes a serine/threonine protein kinase. Although this gene product is similar to serum- and glucocorticoid-induced protein kinase (SGK), this gene is not induced by serum or glucocorticoids. This gene is induced in response to signals that activate phosphatidylinositol 3-kinase, which is also true for SGK. Two alternate transcripts encoding two different isoforms have been described. (provided by RefSeq)

Immunogen

SGK2 (AAH65511, 293 a.a. ~ 367 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
SPINWDDLYHKRLTPPFNPNVTGPADLKHFDPEFTQEAVSKSIGCTPDTVASSSGASSAFLGFSYAPEDDDILDC

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service