Skip to Content
Merck
All Photos(4)

Key Documents

SAB1408048

Sigma-Aldrich

Anti-C1ORF25 antibody produced in mouse

purified immunoglobulin, buffered aqueous solution

Synonym(s):

MGC25112, MGC57134, MST070, MSTP070, TRM1L, bG120K12.3

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
R$4,261.00

R$4,261.00


Estimated to ship onMay 26, 2025



Select a Size

Change View
50 μG
R$4,261.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

R$4,261.00


Estimated to ship onMay 26, 2025


biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~81.7 kDa

species reactivity

human

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
indirect immunofluorescence: suitable
western blot: 1 μg/mL

NCBI accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... C1orf25(81627)

Related Categories

General description

This gene encodes a protein that has some similarity to N2,N2-dimethylguanosine tRNA methyltransferase from other organisms. Studies of the mouse ortholog have shown that this protein plays a role in motor coordination and exploratory behavior, and it may also be involved in modulating postnatal neuronal functions. Alternatively spliced transcripts have been identified for this gene, but their full-length nature is not known. (provided by RefSeq)

Immunogen

C1orf25 (AAH45535.1, 1 a.a. ~ 733 a.a) full-length human protein.

Sequence
MENMAEEELLPLEKEEVEVAQVQVPTPARDSAGVPAPAPDSALDSAPTPASAPAPAPALAQAPALSPSLASAPEEAKSKRHISIQRQLADLENLAFVTDGNFDSASSLNSDNLDAGNRQACPLCPKEKFRACNSHKLRRHLQNLHWKVSVEFEGYRMCICHLPCRPVKPNIIGEQITSKMGAHYHCIICSATITRRTDMLGHVRRHMNKGETKSSYIAASTAKPPKEILKEADTDVQVCPNYSIPQKTDSYFNPKMKLNRQLIFCTLAALAEERKPLECLDAFGATGIMGLQWAKHLGNAVKVTINDLNENSVTLIQENCHLNKLKVVVDSKEKEKSDDILEEGEKNLGNIKVTKMDANVLMHLRSFDFIHLDPFGTSVNYLDSAFRNIRNLGIVSVTSTDISSLYAKAQHVARRHYGCNIVRTEYYKELAARIVVAAVARAAARCNKGIEVLFAVALEHFVLVVVRVLRGPTSADETAKKIQYLIHCQWCEERIFQKDGNMVEENPYRQLPCNCHGSMPGKTAIELGPLWSSSLFNTGFLKRMLFESLHHGLDDIQTLIKTLIFESECTPQSQFSIHASSNVNKQEENGVFIKTTDDTTTDNYIAQGKRKSNEMITNLGKKQKTDVSTEHPPFYYNIHRHSIKGMNMPKLKKFLCYLSQAGFRVSRTHFDPMGVRTDAPLMQFKSILLKYSTPTYTGGQSESHVQSASEDTVTERVEMSVNDKAEASGCRRW

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service