Skip to Content
Merck
All Photos(1)

Key Documents

SAB1407540

Sigma-Aldrich

Anti-KCNK10 antibody produced in mouse

purified immunoglobulin, buffered aqueous solution

Synonym(s):

FLJ43399, K2p10.1, TREK-2, TREK2

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
R$4,261.00

R$4,261.00


Estimated to ship onMay 01, 2025



Select a Size

Change View
50 μG
R$4,261.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

R$4,261.00


Estimated to ship onMay 01, 2025


biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~60.3 kDa

species reactivity

human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... KCNK10(54207)

General description

The protein encoded by this gene belongs to the family of potassium channel proteins containing two pore-forming P domains. This channel is an open rectifier which primarily passes outward current under physiological K+ concentrations, and is stimulated strongly by arachidonic acid and to a lesser degree by membrane stretching, intracellular acidification, and general anaesthetics. Several alternatively spliced transcript variants encoding different isoforms have been identified for this gene. (provided by RefSeq)

Immunogen

KCNK10 (NP_612190.1, 1 a.a. ~ 543 a.a) full-length human protein.

Sequence
MKFPIETPRKQVNWDPKVAVPAAAPVCQPKSATNGQPPAPAPTPTPRLSISSRATVVARMEGTSQGGLQTVMKWKTVVAIFVVVVVYLVTGGLVFRALEQPFESSQKNTIALEKAEFLRDHVCVSPQELETLIQHALDADNAGVSPIGNSSNNSSHWDLGSAFFFAGTVITTIGYGNIAPSTEGGKIFCILYAIFGIPLFGFLLAGIGDQLGTIFGKSIARVEKVFRKKQVSQTKIRVISTILFILAGCIVFVTIPAVIFKYIEGWTALESIYFVVVTLTTVGFGDFVAGGNAGINYREWYKPLVWFWILVGLAYFAAVLSMIGDWLRVLSKKTKEEVGEIKAHAAEWKANVTAEFRETRRRLSVEIHDKLQRAATIRSMERRRLGLDQRAHSLDMLSPEKRSVFAALDTGRFKASSQESINNRPNNLRLKGPEQLNKHGQGASEDNIINKFGSTSRLTKRKNKDLKKTLPEDVQKIYKTFRNYSLDEEKKEEETEKMCNSDNSSTAMLTDCIQQHAELENGMIPTDTKDREPENNSLLEDRN

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yosef Avchalumov et al.
Neuropharmacology, 185, 108438-108438 (2020-12-18)
Alterations in the function of prefrontal cortex (PFC) and hippocampus have been implicated in underlying the relapse to alcohol seeking behaviors in humans and animal models of moderate to severe alcohol use disorders (AUD). Here we used chronic intermittent ethanol

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service