Skip to Content
Merck
All Photos(1)

Key Documents

SAB1406998

Sigma-Aldrich

Anti-ZNF238 antibody produced in mouse

purified immunoglobulin, buffered aqueous solution

Synonym(s):

C2H2-171, RP58, TAZ-1, ZBTB18

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
R$4,261.00

R$4,261.00


Check Cart for Availability


Select a Size

Change View
50 μG
R$4,261.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

R$4,261.00


Check Cart for Availability

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~58.4 kDa

species reactivity

human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ZNF238(10472)

General description

C2H2-type zinc finger proteins, such as ZNF238, act on the molecular level as transcriptional activators or repressors and are involved in chromatin assembly.[supplied by OMIM

Immunogen

ZNF238 (NP_006343.2, 1 a.a. ~ 522 a.a) full-length human protein.

Sequence
MEFPDHSRHLLQCLSEQRHQGFLCDCTVLVGDAQFRAHRAVLASCSMYFHLFYKDQLDKRDIVHLNSDIVTAPAFALLLEFMYEGKLQFKDLPIEDVLAAASYLHMYDIVKVCKKKLKEKATTEADSTKKEEDASSCSDKVESLSDGSSHIAGDLPSDEDEGEDEKLNILPSKRDLAAEPGNMWMRLPSDSAGIPQAGGEAEPHATAAGKTVASPCSSTESLSQRSVTSVRDSADVDCVLDLSVKSSLSGVENLNSSYFSSQDVLRSNLVQVKVEKEASCDESDVGTNDYDMEHSTVKESVSTNNRVQYEPAHLAPLREDSVLRELDREDKASDDEMMTPESERVQVEGGMESSLLPYVSNILSPAGQIFMCPLCNKVFPSPHILQIHLSTHFREQDGIRSKPAADVNVPTCSLCGKTFSCMYTLKRHERTHSGEKPYTCTQCGKSFQYSHNLSRHAVVHTREKPHACKWCERRFTQSGDLYRHIRKFHCELVNSLSVKSEALSLPTVRDWTLEDSSQELWK

Physical form

Solution in phosphate buffered saline, pH 7.4

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anie P Masilamani et al.
iScience, 25(7), 104625-104625 (2022-07-09)
Proteolytic cleavage is an important post-translational mechanism to increase protein variability and functionality. In cancer, this process can be deregulated to shut off tumor-suppressive functions. Here, we report that in glioblastoma (GBM), the tumor suppressor ZBTB18 is targeted for protein

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service