Skip to Content
Merck
All Photos(3)

Key Documents

SAB1403498

Sigma-Aldrich

Monoclonal Anti-ANKRD37 antibody produced in mouse

clone 6E8, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Lrp2bp, MGC111507

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
R$4,261.00

R$4,261.00


Check Cart for Availability


Select a Size

Change View
100 μG
R$4,261.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

R$4,261.00


Check Cart for Availability

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

6E8, monoclonal

form

buffered aqueous solution

mol wt

antigen ~43.9 kDa

species reactivity

human

technique(s)

capture ELISA: suitable
immunofluorescence: suitable
indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

Ankyrin repeat domain 37 (ANKRD37) is a protein containing ankyrin repeats (ARs) and a nuclear localization signal (NLS). The protein is expressed in human testis, small intestine, colon, blood leukocytes and human pancreatic adenocarcinoma cells. The gene is located on human chromosome 4q35.1.

Immunogen

ANKRD37 (NP_859077.1, 1 a.a. ~ 158 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
MLLLDCNPEVDGLKHLLETGASVNAPPDPCKQSPVHLAAGSGLACFLLWQLQTGADLNQQDVLGEAPLHKAAKVGSLECLSLLVASDAQIDLCNKNGQTAEDLAWSCGFPDCAKFLTTIKCMQTIKASEHPDRNDCVAVLRQKRSLGSVENTSGKRKC

Biochem/physiol Actions

Ankyrin repeat domain 37 (ANKRD37) is important in protein-protein interactions and signalling pathways. It acts as a target gene for hypoxia-inducible factor 1 (HIF-1).

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

An integrative genomics approach identifies Hypoxia Inducible Factor-1 (HIF-1)-target genes that form the core response to hypoxia
Benita Y, et al.
Nucleic Acids Research, 37(14), 4587-4602 (2009)
Molecular cloning and characterization of a novel human gene containing 4 ankyrin repeat domains
Li J, et al.
Cellular & Molecular Biology Letters, 10(1), 185-193 (2005)
Analysis of loss of heterozygosity on chromosome 4q in hepatocellular carcinoma using high-throughput SNP array
Zhang H, et al.
Oncology Reports, 23(2), 445-455 (2010)
Mouse Fem1b interacts with and induces ubiquitin-mediated degradation of Ankrd37
Shi YQ, et al.
Gene, 485(2), 153-159 (2011)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service