Skip to Content
Merck
All Photos(2)

Key Documents

SAB1403232

Sigma-Aldrich

Monoclonal Anti-ASB3 antibody produced in mouse

clone 3E5, purified immunoglobulin, buffered aqueous solution

Synonym(s):

ASB-3, FLJ10123, FLJ10421, MGC12531, MGC132002, MGC996

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
R$4,261.00

R$4,261.00


Check Cart for Availability


Select a Size

Change View
100 μG
R$4,261.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

R$4,261.00


Check Cart for Availability

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

3E5, monoclonal

form

buffered aqueous solution

mol wt

antigen ~83.09 kDa

species reactivity

human

technique(s)

capture ELISA: suitable
immunofluorescence: suitable
indirect ELISA: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ASB3(51130)

General description

The protein encoded by this gene is a member of the ankyrin repeat and SOCS box-containing (ASB) family of proteins. They contain ankyrin repeat sequence and SOCS box domain. The SOCS box serves to couple suppressor of cytokine signalling (SOCS) proteins and their binding partners with the elongin B and C complex, possibly targeting them for degradation. Multiple alternatively spliced transcript variants have been described for this gene but some of the full length sequences are not known. (provided by RefSeq)

Immunogen

ASB3 (AAH06488.1, 1 a.a. ~ 518 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
MDFTEAYADTCSTVGLAAREGNVKVLRKLLKKGRSVDVADNRGWMPIHEAAYHNSVECLQMLINADSSENYIKMKTFEGFCALHLAASQGHWKIVQILLEAGADPNATTLEETTPLFLAVENGQIDVLRLLLQHGANVNGSHSMCGWNSLHQASFQENAEIIKLLLRKGANKECQDDFGITPLFVAAQYGKLESLSILISSGANVNCQALDKATPLFIAAQEGHTKCVELLLSSGADPDLYCNEDSWQLPIHAAAQMGHTKILDLLIPLTNRACDTGLNKVSPVYSAVFGGHEDCLEILLRNGYSPDAQACLVFGFSSPVCMAFQKDCEFFGIVNILLKYGAQINELHLAYCLKYEKFSIFRYFLRKGCSLGPWNHIYEFVNHAIKAQAKYKEWLPHLLVAGFDPLILLCNSWIDSVSIDTPIFTLEFTNWKTLAPAVERMLSARASNAWILQQHIATVPSLTHLCRLEIRSSLKSERLRSDSYISQLPLPRSLHNYLLYEDVLRMYEVPELAAIQDG

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service