Skip to Content
Merck
All Photos(1)

Key Documents

SAB1403012

Sigma-Aldrich

Monoclonal Anti-CLDN12 antibody produced in mouse

clone 2D8, purified immunoglobulin, buffered aqueous solution

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
R$4,261.00

R$4,261.00


Check Cart for Availability


Select a Size

Change View
100 μG
R$4,261.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

R$4,261.00


Check Cart for Availability

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

2D8, monoclonal

form

buffered aqueous solution

mol wt

antigen ~52.95 kDa

species reactivity

human

technique(s)

capture ELISA: suitable
indirect ELISA: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CLDN12(9069)

General description

Claudins, such as CLDN12, are components of epithelial cell tight junctions. Tight junctions regulate movement of solutes and ions through the paracellular space and prevent mixing of proteins and lipids in the outer leaflet of the apical and basolateral plasma membrane domains (Acharya et al., 2004).[supplied by OMIM

Immunogen

CLDN12 (AAH36754.1, 1 a.a. ~ 244 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
MGCRDVHAATVLSFLCGIASVAGLFAGTLLPNWRKLRLITFNRNEKNLTVYTGLWVKCARYDGSSDCLMYDTTWYSSVDQLDLRVLQFALPLSMLIAMGALLLCLIGMCNTAFRSSVPNIKLAKCLVNSAGCHLVAGLLFFLAGTVSLSPSIWVIFYNIHLNKKFEPVFSFDYAVYVTIASAGGLFMTSLILFIWYCTCKSLPSPFWQPLYSHPPSMHTYSQPYSARSRLSAIEIDIPVVSHTT

Physical form

Solution in phosphate buffered saline, pH 7.4

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Desislava Kolchakova et al.
Biomolecules, 11(5) (2021-05-01)
Claudins are important components of the tight junctions determining barrier properties, cell polarity, and paracellular permeability. Although many functions of claudins in cancer cells have not been elucidated, recent studies have shown that claudins play an important role in cell

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service