Skip to Content
Merck
All Photos(2)

Key Documents

SAB1400817

Sigma-Aldrich

Anti-MOBKL2C antibody produced in mouse

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-MGC26743, Anti-MOB3C

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
R$4,090.00

R$4,090.00


Check Cart for Availability


Select a Size

Change View
50 μG
R$4,090.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

R$4,090.00


Check Cart for Availability

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

western blot: 1 μg/mL

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

The protein encoded by this gene is similar to the yeast Mob1 protein. Yeast Mob1 binds Mps1p, a protein kinase essential for spindle pole body duplication and mitotic checkpoint regulation. Alternatively spliced transcript variants encoding distinct isoforms have been observed. (provided by RefSeq)

Immunogen

MOBKL2C (NP_958805.1, 1 a.a. ~ 216 a.a) full-length human protein.

Sequence
MALCLKQVFAKDKTFRPRKRFEPGTQRFELYKKAQASLKSGLDLRSVVRLPPGENIDDWIAVHVVDFFNRINLIYGTMAERCSETSCPVMAGGPRYEYRWQDERQYRRPAKLSAPRYMALLMDWIEGLINDEEVFPTRVGVPFPKNFQQVCTKILTRLFRVFVHVYIHHFDSILSMGAEAHVNTCYKHFYYFIREFSLVDQRELEPLREMTERICH

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service