SAB1306738
ANTI-GMFG antibody produced in mouse
crude ascites, buffered aqueous solution
Synonym(s):
GMFG, Glia maturation factor gamma
About This Item
biological source
mouse
Quality Level
antibody form
crude ascites
antibody product type
primary antibodies
clone
monoclonal
form
buffered aqueous solution
mol wt
16801 Da
species reactivity
human
technique(s)
western blot: 1:40000
isotype
IgG1
NCBI accession no.
UniProt accession no.
shipped in
wet ice
storage temp.
−20°C
Gene Information
human ... GMFG(9535)
Physical form
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
10 - Combustible liquids
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service