Skip to Content
Merck
All Photos(1)

Key Documents

SAB1305074

Sigma-Aldrich

MONOCLONAL ANTI-FSHB/FSH (CENTER) antibody produced in mouse

clone 365CT24.1.1, IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

FSHB, Follicle-stimulating hormone beta subunit, Follitropin beta chain, Follitropin subunit beta

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

mouse

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

365CT24.1.1, monoclonal

form

buffered aqueous solution

mol wt

14700 Da

species reactivity

human

technique(s)

western blot: 1:150-1:1500

isotype

IgG3

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... FSHB(2488)

Physical form

Supplied in PBS with 0.09% (W/V) sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service