Skip to Content
Merck
All Photos(1)

Key Documents

S8524

Sigma-Aldrich

N-Succinyl-Ile-Ile-Trp-7-amido-4-methylcoumarin

protease substrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C37H45N5O8
CAS Number:
Molecular Weight:
687.78
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:

form

solid

storage temp.

−20°C

SMILES string

CC[C@H](C)[C@H](NC(=O)CCC(O)=O)C(=O)N[C@@H]([C@@H](C)CC)C(=O)N[C@@H](Cc1c[nH]c2ccccc12)C(=O)Nc3ccc4C(C)=CC(=O)Oc4c3

InChI

1S/C37H45N5O8/c1-6-20(3)33(41-30(43)14-15-31(44)45)37(49)42-34(21(4)7-2)36(48)40-28(17-23-19-38-27-11-9-8-10-26(23)27)35(47)39-24-12-13-25-22(5)16-32(46)50-29(25)18-24/h8-13,16,18-21,28,33-34,38H,6-7,14-15,17H2,1-5H3,(H,39,47)(H,40,48)(H,41,43)(H,42,49)(H,44,45)/t20-,21-,28-,33-,34-/m0/s1

InChI key

DIQQFBVJYYDILY-DFXXCKAUSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G C Kundu et al.
Life sciences, 50(13), 965-970 (1992-01-01)
An enzyme partially purified from bovine lung membranes appears to be endothelin converting enzyme (ECE). This enzyme specifically cleaves big endothelin-1 (big ET-1) at the proper site, between Trp21 and Val22, with maximum activity at pH 7.5 and with a
G C Kundu et al.
International journal of peptide and protein research, 42(1), 64-67 (1993-07-01)
The metal ion of endothelin-converting enzyme (ECE) was investigated by inhibiting the enzyme with ethylenediaminetetraacetic acid (EDTA) and restoring activity by adding divalent metal salts in quantities less than the concentration of EDTA. Under these conditions, only metal ions that
Salvatore de Stefano et al.
Natural product letters, 16(3), 207-211 (2002-06-07)
A novel metabolite related to the previously identified compound 3-O-methylfunicone (1) was isolated from the fungus Penicillium pinophilum and its structure elucidated by spectral analysis as 3-O-methyl-5, 6-epoxyfunicone (2).
Endothelin converting enzyme of porcine lung.
L J Murphy et al.
Biochemical Society transactions, 21(1), 27S-27S (1993-02-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service