Skip to Content
Merck
All Photos(1)

Key Documents

S8508

Sigma-Aldrich

[Tyr11]-Somatostatin

≥90% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C76H104N18O20S2
CAS Number:
Molecular Weight:
1653.88
MDL number:
UNSPSC Code:
12352200

Assay

≥90% (HPLC)

storage temp.

−20°C

SMILES string

CC(N)C(=O)NCC(=O)NC1CSSCC(NC(=O)C(CO)NC(=O)C(NC(=O)C(Cc2ccc(O)cc2)NC(=O)C(NC(=O)C(CCCCN)NC(=O)C(Cc3c[nH]c4ccccc34)NC(=O)C(Cc5ccccc5)NC(=O)C(Cc6ccccc6)NC(=O)C(CC(N)=O)NC(=O)C(CCCCN)NC1=O)C(C)O)C(C)O)C(O)=O

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S J Sullivan et al.
Endocrinology, 122(3), 1137-1145 (1988-03-01)
Previous studies with heterogeneous populations of pancreatic cells have provided evidence for the presence of somatostatin (SRIF) receptors in cytosol and secretion vesicles, as well as the plasma membrane. To examine the distribution of SRIF receptors between soluble and membrane
B Colás et al.
Journal of receptor research, 7(5), 653-666 (1987-01-01)
Specific binding sites for somatostatin have been detected in cytosolic fraction of bovine cystic duct mucosa. At 37 degrees C, the interaction of 125I-Tyr11-somatostatin with cytosolic fraction was rapid, reversible, specific and saturable. At equilibrium, the binding of tracer was
A Schonbrunn et al.
Endocrinology, 132(1), 146-154 (1993-01-01)
The neuropeptide somatostatin (SRIF) triggers its biological effects by binding to high affinity membrane receptors. To develop a ligand useful for receptor affinity purification and localization, we have examined the ability of a novel monobiotinylated SRIF derivative to bind to
R Tatoud et al.
Molecular and cellular endocrinology, 113(2), 195-204 (1995-09-22)
We investigated somatostatin receptors (SSTRs) in surgical specimens of prostate cancer and benign prostate hyperplasia (BPH), a normal immortalized epithelial cell line (PNT1), epithelial cancer cell lines, and stromal cells in short-term culture derived from normal and BPH biopsies. Cross-linking
Effects of urethane administration on somatostatin receptors in rat cerebral cortex and pituitary.
H Iwahana et al.
The Tokushima journal of experimental medicine, 35(3-4), 69-77 (1988-12-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service