Skip to Content
Merck
All Photos(1)

Key Documents

S8259

Sigma-Aldrich

2,2′-Sulfinyl-bis(4,6-dichlorophenol)

Synonym(s):

Bithionol sulfoxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C12H6O3Cl4S
CAS Number:
Molecular Weight:
372.05
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

SMILES string

Oc1c(Cl)cc(Cl)cc1S(=O)c2cc(Cl)cc(Cl)c2O

InChI

1S/C12H6Cl4O3S/c13-5-1-7(15)11(17)9(3-5)20(19)10-4-6(14)2-8(16)12(10)18/h1-4,17-18H

InChI key

RPAJWWXZIQJVJF-UHFFFAOYSA-N

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E Chroustová et al.
Veterinarni medicina, 25(9), 557-563 (1980-09-01)
The elimination of the sources of invasion, i. e. hay and forage coming from the infested biotopes, combined with the administration of bithionol sulphoxide, resulted in a reduction of the invasion of cattle by Fascioloides magna from 21.1 to 3.2%
D Mourot et al.
Mutation research, 188(1), 53-55 (1987-05-01)
The mutagenic effects of bithionol sulfoxide and its two major metabolites, bithionol and bithionol sulfone, on 4 Salmonella typhimurium strains (TA97, TA98, TA100 and TA102) were investigated. Bithionol sulfoxide was found to be mutagenic to TA98 and TA100. However, mutagenicity
D Mourot et al.
Journal - Association of Official Analytical Chemists, 70(5), 810-812 (1987-09-01)
A liquid chromatographic method is described for determining bithionol sulfoxide and its metabolites, bithionol and bithionol sulfone, in milk. Samples are treated with HCl to precipitate proteins and to permit extraction of bithionol sulfoxide in nonionized form. Tetrahydrofuran is added
M Dagorn
Toxicological European research. Recherche europeenne en toxicologie, 4(2), 101-105 (1982-03-01)
The distribution of a single oral dose of 14C bithionol sulfoxide in mice was determined by whole body autoradiography. The bithionol sulfoxide was found in the whole organism mainly in richly vascularized organs, liver, kidney, lung. It did not pass
[Ionization constants of bithionol sulfoxide].
M Dagorn et al.
Annales pharmaceutiques francaises, 41(5), 473-481 (1983-01-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service