Skip to Content
Merck
All Photos(1)

Key Documents

S7029

Sigma-Aldrich

[Pro9]-Substance P

≥97% (HPLC)

Synonym(s):

Arg-Pro-Lys-Pro-Gln-Gln-Phe-Phe-Pro-Leu-Met-NH2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C66H102N18O13S
CAS Number:
Molecular Weight:
1387.69
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥97% (HPLC)

storage temp.

−20°C

SMILES string

CSCCC(NC(=O)C(CC(C)C)NC(=O)C1CCCN1C(=O)C(Cc2ccccc2)NC(=O)C(Cc3ccccc3)NC(=O)C(CCC(N)=O)NC(=O)C(CCC(N)=O)NC(=O)C4CCCN4C(=O)C(CCCCN)NC(=O)C5CCCN5C(=O)C(N)CCCNC(N)=N)C(N)=O

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Y Torrens et al.
Neuropeptides, 31(3), 243-251 (1997-06-01)
The rat urinary bladder possesses NK1, NK2 (but not NK3) and 'septide-sensitive' tachykinin receptors coupled to a phospholipase C. The present study performed with SR48968 (10(-6) M) to avoid any interaction of the tested peptides with NK2 receptors, indicates that
C Gauchy et al.
Neuroscience, 73(4), 929-939 (1996-08-01)
The effects of septide (a short substance P C-terminal analogue) and of the neurokinin-1 receptor agonist [Pro9]substance P on the N-methyl-D-aspartate (50 microM)-evoked release of [3H]dopamine (continuously synthesized from [3H]tyrosine) were investigated in the absence or the presence of the
S Lavielle et al.
European journal of pharmacology, 258(3), 273-276 (1994-06-13)
In the guinea-pig ileum tissue, [Pro9]substance P, a tachykinin NK1 receptor selective agonist and septide, [pGlu6,Pro9]-substance P-(6-11), do not interact with the same receptor as shown by the different inhibitory profiles of GR 72251 and [D-Pro9,Pro10,Trp11]substance P. Substitution at position
J C Delumeau et al.
Journal of neurochemistry, 57(6), 2026-2035 (1991-12-01)
The effects on cytosolic Ca2+ concentration of 2-chloroadenosine and [L-Pro9]-substance P, a selective agonist of NK1 receptors, were investigated on astrocytes from embryonic mice in primary culture. Cells responded to [L-Pro9]-substance P with a transitory increase in cytosolic Ca2+ which
F Petitet et al.
Journal of neurochemistry, 56(3), 879-889 (1991-03-01)
Previous studies have indicated that [Pro9]-substance P ([Pro9]-SP) possesses very good affinity for NK-1 binding sites and that, in contrast to substance P, it interacts selectively with these sites. Therefore, [3H][Pro9]-SP (75 Ci/mmol) was synthesized in order to study its

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service