Skip to Content
Merck
All Photos(1)

Key Documents

S6879

Sigma-Aldrich

D-Sphingosine

from bovine brain, ~99% (TLC)

Synonym(s):

(2S,3R,4E)-2-Amino-4-octadecene-1,3-diol, 4-Sphingenine, trans-D-erythro-2-Amino-4-octadecene-1,3-diol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H37NO2
CAS Number:
Molecular Weight:
299.49
Beilstein:
1727294
EC Number:
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:

biological source

bovine brain

Assay

~99% (TLC)

storage temp.

−20°C

SMILES string

OC[C@@](N)([H])[C@]([H])(O)/C=C/CCCCCCCCCCCCC

InChI

1S/C18H37NO2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-18(21)17(19)16-20/h14-15,17-18,20-21H,2-13,16,19H2,1H3/b15-14+/t17-,18+/m0/s1

InChI key

WWUZIQQURGPMPG-KRWOKUGFSA-N

Looking for similar products? Visit Product Comparison Guide

Application

D-Sphingosine from bovine brain was tested for effects on distribution of clathrin/AP-2 in cultured fibroblasts.8 It was used to prepare vesicles and study the interactions of DNA with bilayers of cationic liposomes.9

Biochem/physiol Actions

A constituent of cell membranes. Precursor of ceramide. Selective inhibitor of protein kinase C, but does not inhibit protein kinase A or myosin light chain kinase. Inhibitor of calmodulin-dependent enzymes.

Quality

May contain small amounts of structural analogues.

Preparation Note

Prepared from sphingomyelin.

comparable product

Product No.
Description
Pricing

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 4

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Susan M Hancock et al.
Nature chemical biology, 5(7), 508-514 (2009-06-16)
Though glycosphingolipids have great potential as therapeutics for cancer, HIV, neurodegenerative diseases and auto-immune diseases, both extensive study of their biological roles and development as pharmaceuticals are limited by difficulties in their synthesis, especially on large scales. Here we addressed
Meret E Ricklin et al.
Neurology, 81(2), 174-181 (2013-05-24)
To study the immune response against varicella-zoster virus (VZV) in patients with multiple sclerosis before and during fingolimod therapy. The VZV-specific immune response was studied using interferon (IFN)-γ enzyme-linked immunosorbent spot assay, proliferation assays, and upregulation of T-cell activation markers
Tim Ting Chiu et al.
The Journal of biological chemistry, 288(24), 17520-17531 (2013-05-04)
Insulin activates a cascade of signaling molecules, including Rac-1, Akt, and AS160, to promote the net gain of glucose transporter 4 (GLUT4) at the plasma membrane of muscle cells. Interestingly, constitutively active Rac-1 expression results in a hormone-independent increase in
Jeroen Tibboel et al.
Chest, 145(1), 120-128 (2014-01-08)
Sphingolipids comprise a class of bioactive lipids that are involved in a variety of pathophysiologic processes, including cell death and survival. Ceramide and sphingosine-1-phosphate (S1P) form the center of sphingolipid metabolism and determine proapoptotic and antiapoptotic balance. Findings in animal
David Henault et al.
Neurology, 81(20), 1768-1772 (2013-10-18)
To determine the range of fluctuation in total lymphocyte counts (TLCs) in peripheral blood over a 4- to 7-year period in patients with MS receiving fingolimod (FTY720) and the relation between TLCs and T-cell subsets (CD4+, CD8+, CCR7+/-) that are

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service