Skip to Content
Merck
All Photos(2)

Documents

S6446

Sigma-Aldrich

Anti-SMC1L1 (C-terminal) antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-SMC1, Anti-SMC1 structural maintenance of chromosomes 1-like 1, Anti-SMC1α

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 160 kDa

species reactivity

rat (predicted), human, mouse (predicted)

technique(s)

immunoprecipitation (IP): 5-10 μL using HEK 293-T cell lysates
indirect immunofluorescence: 1:100-1:200 using paraformaldehyde-fixed HEK 293-T cells
microarray: suitable
western blot: 1:500-1:1,000 using HeLa nuclear extracts

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SMC1A(8243)
mouse ... Smc1a(24061)
rat ... Smc1a(63996)

General description

Structural maintenance of chromosomes protein 1A (SMC1A, SMC1L1, SMC1), a structural maintenance of chromosomes (SMC) protein belongs to the SMC superfamily and is an important constituent of the cohesin complex. This gene is located on human chromosome Xp11.22.

Specificity

Anti SMC1L1 (also known as SMC1 and SMC1a) specifically recognizes SMC1L1.

Immunogen

synthetic peptide corresponding to amino acids 1210-1225 of human SMC1L1, conjugated to KLH via an N-terminal cysteine residue. The immunizing sequence is conserved in mouse and rat.

Application

Anti-SMC1L1 (C-terminal) antibody produced in rabbit may be used in:
  • immunoblotting
  • immunoprecipitation
  • immunofluorescence

Biochem/physiol Actions

Structural maintenance of chromosomes protein 1A (SMC1A, SMC1L1, SMC1) can be phosphorylated by Ataxia-telangiectasia, mutated (ATM) indicating a potential role for this protein in DNA repair. Thus, SMC1L1 phosphorylation appears to be the critical downstream event in the ATM-nibrin (NBS1)-breast cancer type 1 (BRCA1) pathway, which mediates cell survival and chromosomal stabilization after DNA damage. Mutations in SMC1L1 gene result in the Cornelia de Lange syndrome, a multisystem developmental disorder. It participates in chromosome dynamics, double-strand break (DSB) repair and tumorigenesis. This gene can control the cell cycle and helps to maintain the stability of genome. Suppression of SMC1A can block the proliferation and migration of hepatocellular carcinoma cells.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Storage and Stability

For continuous use, store at 2-8 °C for up to one month. For extended storage, freeze in working aliquots. Repeated freezing and thawing, or storage in “frost-free” freezers, is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilutions should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog, our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

related product

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Phosphorylation of SMC1 is a critical downstream event in the ATM-NBS1-BRCA1 pathway
Kitagawa R, et al.
Genes & Development, 18(12), 1423-1438 (2004)
Phosphorylation of SMC1A promotes hepatocellular carcinoma cell proliferation and migration
Zhang Y, et al.
International Journal of Biological Sciences, 14(9), 1081-1081 (2018)
Ilaria Parenti et al.
Epigenetics, 9(7), 973-979 (2014-04-24)
Cornelia de Lange syndrome (CdLS) is a rare multisystem disorder characterized by facial dysmorphisms, limb anomalies, and growth and cognitive deficits. Mutations in genes encoding subunits (SMC1A, SMC3, RAD21) or regulators (NIPBL, HDAC8) of the cohesin complex account for approximately

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service