Key Documents
S5939
Sigma-Aldrich
Sun cap closures
Biofilter, O.D. 18 mm
About This Item
UNSPSC Code:
41100000
description
disposable
feature
autoclavable
O.D.
18 mm
application(s)
agriculture
General description
8 mm 0.04 μm polypropylene filter disc with heatproof adhesive. Comes with 20 closures per sheet and 25 total sheets.