S5508
Statine
Synonym(s):
(3S,4S)-4-Amino-3-hydroxy-6-methylheptanoic acid
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C8H17NO3
CAS Number:
Molecular Weight:
175.23
Beilstein:
3537971
MDL number:
UNSPSC Code:
12352200
storage temp.
−20°C
SMILES string
CC(C)C[C@H](N)[C@@H](O)CC(O)=O
Looking for similar products? Visit Product Comparison Guide
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service