Skip to Content
Merck
All Photos(1)

Key Documents

S4443

Sigma-Aldrich

SCH-28080

≥98% (HPLC), solid

Synonym(s):

2-methyl-8-(phenylmethoxy)imidazo[1,2-a]pyridine-3-acetonitrile

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
R$2,064.00
50 MG
R$9,241.00

R$2,064.00


Estimated to ship onMay 11, 2025


Request a Bulk Order

Select a Size

Change View
10 MG
R$2,064.00
50 MG
R$9,241.00

About This Item

Empirical Formula (Hill Notation):
C17H15N3O
CAS Number:
Molecular Weight:
277.32
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

R$2,064.00


Estimated to ship onMay 11, 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

solid

color

white to light tan

solubility

DMSO: soluble >10 mg/mL
H2O: insoluble

compatibility

for use with ABI 7700

storage temp.

−20°C

SMILES string

Cc1nc2c(OCc3ccccc3)cccn2c1CC#N

InChI

1S/C17H15N3O/c1-13-15(9-10-18)20-11-5-8-16(17(20)19-13)21-12-14-6-3-2-4-7-14/h2-8,11H,9,12H2,1H3

InChI key

PYKJFEPAUKAXNN-UHFFFAOYSA-N

Application

SCH-28080 was used to treat zebrafish embryos to study the role of H+/K+-ATPase in establishment of left-right axis during development.[1]

Biochem/physiol Actions

SCH-28080 is a potent inhibitor of gastric H+ and K+-ATPase. The novel antiulcer agents, SCH-28080 and SCH-32651 were examined for their ability to inhibit the H+K+ ATPase enzyme activity in a preparation of microsomal membranes from rabbit fundic mucosa. SCH- 28080 inhibited the isolated enzyme activity with a potency similar to omeprazole, IC50s of 2.5 and 4.0 μM respectively. SCH 32651 was less potent exhibiting an IC50 of 200.0 μM. Both compounds may therefore exert their antisecretory activity via a direct inhibition of the parietal cell H+K+ ATPase.
SCH-28080 is a potent inhibitor of gastric H+ and K+-ATPase.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jiahong Shao et al.
Biochimica et biophysica acta, 1800(9), 906-911 (2010-07-03)
The H,K-ATPase, consisting of α and ß subunits, belongs to the P-type ATPase family. There are two isoforms of the α subunit, HKα₁ and HKα₂ encoded by different genes. The ouabain-resistant gastric HKα₁-H,K-ATPase is Sch28080-sensitive. However, the colonic HKα₂-H,K-ATPase from
P Kirchhoff et al.
American journal of physiology. Gastrointestinal and liver physiology, 291(5), G838-G843 (2006-06-27)
The gastric H+,K+-ATPase of the parietal cell is responsible for acid secretion in the stomach and is the main target in the pharmacological treatment of acid-related diseases. Omeprazole and other benzimidazole drugs, although having delayed efficacy if taken orally, have
I Jeanette Lynch et al.
American journal of physiology. Renal physiology, 298(2), F408-F415 (2009-11-20)
In the collecting duct (CD), H-K-ATPases function in cation reabsorption and H secretion. This study evaluated H-K-ATPase-mediated H secretion along the mouse CD, measured as EIPA- and luminal bafilomycin A(1)-insensitive intracellular pH (pH(i)) recovery from acute H loading (NH(4)) using
Taku Hibino et al.
Development genes and evolution, 216(5), 265-276 (2006-03-15)
The degree of conservation among phyla of early mechanisms that pattern the left-right (LR) axis is poorly understood. Larvae of sea urchins exhibit consistently oriented LR asymmetry. The main part of the adult rudiment is formed from the left coelomic
Herman G P Swarts et al.
Biochimica et biophysica acta, 1768(3), 580-589 (2006-12-02)
The primary sequence of non-gastric H,K-ATPase differs much more between species than that of Na,K-ATPase or gastric H,K-ATPase. To investigate whether this causes species-dependent differences in enzymatic properties, we co-expressed the catalytic subunit of human non-gastric H,K-ATPase in Sf9 cells

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service