Skip to Content
Merck
All Photos(1)

Key Documents

S2886

Sigma-Aldrich

[pGlu5]-Substance P Fragment 5-11

≥95% (HPLC)

Synonym(s):

pGlu-Gln-Phe-Phe-Gly-Leu-Met-NH2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C41H57N9O9S
CAS Number:
Molecular Weight:
852.01
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥95% (HPLC)

storage temp.

−20°C

SMILES string

CSCCC(NC(=O)C(CC(C)C)NC(=O)CNC(=O)C(Cc1ccccc1)NC(=O)C(Cc2ccccc2)NC(=O)C(CCC(N)=O)NC(=O)C3CCC(=O)N3)C(N)=O

InChI

1S/C41H57N9O9S/c1-24(2)20-30(40(58)47-27(36(43)54)18-19-60-3)46-35(53)23-44-37(55)31(21-25-10-6-4-7-11-25)49-41(59)32(22-26-12-8-5-9-13-26)50-39(57)29(14-16-33(42)51)48-38(56)28-15-17-34(52)45-28/h4-13,24,27-32H,14-23H2,1-3H3,(H2,42,51)(H2,43,54)(H,44,55)(H,45,52)(H,46,53)(H,47,58)(H,48,56)(H,49,59)(H,50,57)

InChI key

XDUVKSVLDVUFCJ-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R W Bury et al.
Journal of medicinal chemistry, 19(6), 854-856 (1976-06-01)
Substance P (Arg-Pro-Lys-Pro-Gln-Gln-Phe-Phe-Gly-Leu-Met-NH2) and the C-terminal partial sequences down to the tripeptide were synthesized by a solid-phase method. These peptides were assayed for vasodilator, spasmogenic, and venoconstrictor properties using three preparations, viz. the hind limb blood flow of the dog

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service