Skip to Content
Merck
All Photos(1)

Key Documents

S2690

Sigma-Aldrich

Anti-phospho-STAT3 (pTyr705) antibody produced in rabbit

affinity isolated antibody, buffered aqueous glycerol solution

Sign Into View Organizational & Contract Pricing

Select a Size

1 VIAL
R$5,878.00

R$5,878.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 VIAL
R$5,878.00

About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

R$5,878.00


Check Cart for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous glycerol solution

species reactivity

human, rat, mouse

technique(s)

western blot: suitable

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... STAT3(6774)
mouse ... Stat3(20848)
rat ... Stat3(25125)

Immunogen

synthetic phosphopeptide derived from the region of human STAT 3 that contains tyrosine705.

Physical form

Solution in Dulbecco′s phosphate buffered saline (without Mg2+ and Ca2+), pH 7.3(+/-0.1), containing 1 mg/mL BSA (IgG and protease-free), 50% glycerol and 0.05% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bhavnita Soni et al.
Molecules (Basel, Switzerland), 27(1) (2022-01-12)
Macrophage phenotype plays a crucial role in the pathogenesis of Leishmanial infection. Pro-inflammatory cytokines signals through the Janus kinase/signal transducer and activator of transcription (JAK/STAT) pathway that functions in parasite killing. Suppression of cytokine signaling (SOCS) is a well-known negative
Xiu-Fen Yang et al.
Chinese medical journal, 131(1), 75-81 (2017-12-23)
Leukemia inhibitory factor (LIF) has been reported to possess various pharmacological effects, including displaying vascular and neuroprotective properties, during retinal disease. The aim of this study was to investigate the vascular and structural changes in the retina of diabetic mice
Duanqun He et al.
Folia histochemica et cytobiologica, 58(2), 127-134 (2020-06-20)
Oxidative stress and cell apoptosis have both been suggested to be closely associated with the pathogenesis of Parkinson's disease (PD). Previously, bisdemethoxycurcumin (BDMC) has been shown to exhibit several desirable characteristics as a candidate neuroprotective agent, including antioxidant and anti-inflammatory
Tanja Babic et al.
American journal of physiology. Regulatory, integrative and comparative physiology, 296(4), R845-R854 (2009-01-30)
The satiating potency of CCK has been well characterized, including its mediation by capsaicin-sensitive vagal primary afferents. We have previously shown that peripherally administered CCK activates the MAPK-signaling cascade in a population of nucleus of the solitary tract (NTS) neurons

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service