S1753
D-Sorbitol 6-phosphate barium salt
≥90%
Synonym(s):
D-Glucitol 6-phosphate
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Linear Formula:
C6H13O9PBa
CAS Number:
Molecular Weight:
397.46
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25
Recommended Products
Assay
≥90%
form
solid
impurities
water (Karl Fischer, NMT 8.5% (2 MOLES/MOLE))
color
white
storage temp.
−20°C
SMILES string
[Ba++].OC[C@H](O)[C@@H](O)[C@H](O)[C@H](O)COP([O-])([O-])=O
InChI
1S/C6H15O9P.Ba/c7-1-3(8)5(10)6(11)4(9)2-15-16(12,13)14;/h3-11H,1-2H2,(H2,12,13,14);/q;+2/p-2/t3-,4+,5+,6+;/m0./s1
InChI key
ANEIDNTXSPRLJW-BTVCFUMJSA-L
Other Notes
To gain a comprehensive understanding of our extensive range of Sugar alcohols for your research, we encourage you to visit our Carbohydrates Category page.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Inhalation - Acute Tox. 4 Oral
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service