Skip to Content
Merck
All Photos(1)

Key Documents

RABCTYROD

Sigma-Aldrich

Fixing Solution for Phosphotyrosine Assay (Item D)

Sign Into View Organizational & Contract Pricing

Select a Size

30 ML
R$228.00

R$228.00


Check Cart for Availability


Select a Size

Change View
30 ML
R$228.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

R$228.00


Check Cart for Availability

shipped in

wet ice

storage temp.

−20°C

Other Notes

This item is for RAB series CBEL ELISA kits.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

68.0 °F - closed cup

Flash Point(C)

20 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service