Skip to Content
Merck
All Photos(1)

Key Documents

RAB1656

Sigma-Aldrich

Human Pleiotrophin ELISA

for serum, plasma and cell culture supernatants

Sign Into View Organizational & Contract Pricing

Select a Size

96 WELLS
R$5,181.00

R$5,181.00


Estimated to ship onMay 21, 2025



Select a Size

Change View
96 WELLS
R$5,181.00

About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

R$5,181.00


Estimated to ship onMay 21, 2025


species reactivity

human

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable

input

sample type plasma
sample type cell culture supernatant(s)
sample type serum

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 1.3 ng/mL
standard curve range: 1.229-300 ng/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... PTN(5764)

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached Protocolfor details.

Other Notes

A sample Certificate of Analysis is available for this product. Please type the word sample in the text box provided for lot number.

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service