Skip to Content
Merck
All Photos(1)

Key Documents

RAB1284

Sigma-Aldrich

Human BAMBI ELISA

for serum, plasma and cell culture supernatants

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.32
Pricing and availability is not currently available.

species reactivity

human

technique(s)

ELISA: suitable

input

sample type plasma
sample type serum
sample type cell culture supernatant(s)

assay range

inter-assay cv: <12%
intra-assay cv: <10%

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... BAMBI(25805)

General description

BAMBI (bone morphogenic protein and activin membrane-bound inhibitor) gene codes for a transmembrane glycoprotein which is a pseudo receptor belonging to the family of TGF-β (transforming growth factor β receptor) type I receptors.[1][2] BAMBI is predominantly expressed in placenta, spleen and kidney medulla and also found liver, prostate, gut and kidney cortex.[1]
This ELISA antibody pair detects Human BAMBI. Other species not yet determined.

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached Protocolfor details.

Biochem/physiol Actions

BAMBI (bone morphogenic protein and activin membrane-bound inhibitor) acts as an inhibitor of TGF-β (transforming growth factor β receptor) signalling which causes epithelial-mesenchymal transition.[1] Upregulation of BAMBI gene is observed in chronic obstructive pulmonary disease and a number of cancer including thyroid, liver, colorectal cancer and ovarian cancer.[3][4] BAMBI might regulate the events of inflammation and remodeling processes.[3]

Other Notes

A sample Certificate of Analysis is available for this product. Please type the word sample in the text box provided for lot number.

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Downregulation of the TGFβ Pseudoreceptor BAMBI in Non?Small Cell Lung Cancer Enhances TGFβ Signaling and Invasion.
Marwitz S, et al.
Cancer Research (2016)
Dysregulation of the Transforming Growth Factor β Pathway in Induced Pluripotent Stem Cells Generated from Patients with Diamond Blackfan Anemia
Jingping G, et al.
PLoS ONE, 10(8), e0134878-e0134878 (2015)
LPS/TLR4 Signaling Enhances TGF-β Response Through Downregulating BAMBI During Prostatic Hyperplasia
Yao H, et al.
Scientific Reports, 6, 27051-27051 (2016)
Kai Liu et al.
Molecular medicine reports, 10(2), 874-880 (2014-06-11)
The upregulation of bone morphogenetic protein and activin membrane‑bound inhibitor (BAMBI) has been observed in several types of malignant cancer, including thyroid, ovarian, liver and colorectal cancer. However, the pathological role and the regulatory mechanism of BAMBI in gastric cancer

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service