Skip to Content
Merck
All Photos(1)

Documents

RAB1096

Sigma-Aldrich

Human VEGFR1 ELISA Kit

for cell culture supernatants, plasma, and serum samples

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

species reactivity

human

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable

input

sample type cell culture supernatant(s)
sample type serum sample(s)
sample type plasma

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 1.5 ng/ml

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... FLT1(2321)

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached Protocolfor details.

Other Notes

A sample Certificate of Analysis is available for this product. Please type the word sample in the text box provided for lot number.

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhao Zhang et al.
Current medical science, 40(4), 671-676 (2020-08-31)
Acute respiratory distress syndrome (ARDS) is associated with a mortality of 45%. Our previous research indicated that anti-vascular endothelial growth factor (VEGF) could maintain the normal structure and function of the respiratory barrier. However, systemic application of VEGF antagonists would
ELABELA concentration is not decreased in maternal plasma before the onset of preeclampsia.
Patricia Villie et al.
American journal of obstetrics and gynecology, 220(3), 284-285 (2018-12-24)
Elizabeth F Sutton et al.
American journal of physiology. Regulatory, integrative and comparative physiology, 318(6), R1047-R1057 (2020-05-07)
Preeclampsia is a spontaneously occurring, pregnancy-specific syndrome that is clinically diagnosed by new onset hypertension and proteinuria. Epidemiological evidence describes an association between a history of preeclampsia and increased risk for cardiovascular disease in later life; however, the mechanism(s) driving
Haley Smalley et al.
Cancer letters, 495, 66-75 (2020-09-07)
Melanoma, a form of skin cancer, is one of the most common cancers in young men and women. Tumors require angiogenesis to provide oxygen and nutrients for growth. Pro-angiogenic molecules such as VEGF and anti-angiogenic molecules such as sFlt-1 control
Lisa A Vrooman et al.
Development (Cambridge, England), 147(11) (2020-05-31)
Although widely used, assisted reproductive technologies (ARTs) are associated with adverse perinatal outcomes. To elucidate their underlying causes, we have conducted a longitudinal analysis of placental development and fetal growth using a mouse model to investigate the effects of individual

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service