Skip to Content
Merck
All Photos(1)

Key Documents

RAB0719

Sigma-Aldrich

Human SSSCA1 / Sjoegren Syndrome/Scleroderma Autoantigen 1 ELISA Kit

Sign Into View Organizational & Contract Pricing

Select a Size

96 WELLS
R$5,181.00

R$5,181.00


Estimated to ship onJune 02, 2025



Select a Size

Change View
96 WELLS
R$5,181.00

About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

R$5,181.00


Estimated to ship onJune 02, 2025


species reactivity

human

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable

assay range

inter-assay cv: <10%
intra-assay cv: <12%

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... SSSCA1(10534)

General description

The SSSCA1 (Sjoegren syndrome/scleroderma autoantigen 1) gene codes for a cell cycle inhibitor, also called as p27.[1] The SSSCA1 gene is mapped to human chromosome 11q13.1.[2]
The antibody pair provided in this kit recognizes human SSSCA1.

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Biochem/physiol Actions

Sjoegren Syndrome Autoantigen 1 is an important cyclin-dependent kinase complexes inhibitor and prevents the G1 to S phase transition during cell cycle. It is regulated by ubiquitin-proteasome.[1] The Sjoegren Syndrome Autoantigen 1 gene expression is downregulated in cancer and indicates poor prognosis.[3]

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Loss of 3p26. 3 is an independent prognostic factor in patients with oral squamous cell carcinoma.
Uchida K, et al.
Oncology Reports, 26(2), 463-469 (2011)
Subcellular localization of p27 and prostate cancer recurrence: automated digital microscopy analysis of tissue microarrays.
Ananthanarayanan V, et al.
Human Pathology, 42(6), 873-881 (2011)
Expression of CRM1 in human gliomas and its significance in p27 expression and clinical prognosis.
Shen A, et al.
Neurosurgery, 65(1), 153-160 (2009)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service