Skip to Content
Merck
All Photos(1)

Key Documents

RAB0698

Sigma-Aldrich

Human IL24 / Interleukin-24 ELISA Kit

Sign Into View Organizational & Contract Pricing

Select a Size

96 WELLS
R$3,289.00

R$3,289.00


Estimated to ship onMay 26, 2025



Select a Size

Change View
96 WELLS
R$3,289.00

About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

R$3,289.00


Estimated to ship onMay 26, 2025


species reactivity

human

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable

input

sample type cell culture supernatant(s)
sample type plasma
sample type serum

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 0.41 ng/mL
standard curve range: 0.41-100 ng/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... IL24(11009)

General description

The antibody pair provided in this kit recognizes Human Interleukin-24.

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yao Zhang et al.
The protein journal, 38(5), 576-585 (2019-05-08)
Interleukin 24 (IL-24) has a broad spectrum of specific antitumor activities without affecting normal cells. The recombinant human IL-24 (rhIL-24) expressed in E. coli has low biological activity due to lack of necessary glycosylation modification. In this study, based on
Anna Steinert et al.
Journal of immunology (Baltimore, Md. : 1950), 199(7), 2570-2584 (2017-09-03)
IL-19, a member of the IL-10 cytokine family that signals through the IL-20 receptor type I (IL-20Rα:IL-20Rβ), is a cytokine whose function is not completely known. In this article, we show that the expression of
Zheng Wu et al.
Cancer cell international, 20, 33-33 (2020-02-06)
Interleukin-24 (IL-24) is a therapeutic gene for melanoma, which can induce melanoma cell apoptosis. Mesenchymal stem cells (MSCs) show promise as a carrier to delivery anti-cancer factors to tumor tissues. Induced pluripotent stem cells (iPSCs) are an alternative source of
Xiaojing Xu et al.
Molecular therapy oncolytics, 14, 213-221 (2019-07-25)
Interleukin-24 (IL-24) is a promising agent for cancer immunotherapy that induces apoptosis of tumor cells and enhances T cell activation and function. In order to improve the antitumor activity induced by Newcastle disease virus (NDV)-modified tumor vaccine, we generated a recombinant
Johanna C Klein et al.
Nature communications, 8, 14600-14600 (2017-03-17)
Toll-like receptors (TLRs) are located either on the cell surface or intracellularly in endosomes and their activation normally contributes to the induction of protective immune responses. However, in cancer their activation by endogenous ligands can modulate tumour progression. It is

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service