Skip to Content
Merck
All Photos(2)

Documents

RAB0401

Sigma-Aldrich

Human PECAM-1 ELISA Kit

for serum, plasma, cell culture supernatants and urine

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

species reactivity

human

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable
capture ELISA: suitable

input

sample type serum
sample type plasma
sample type cell culture supernatant(s)
sample type urine

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 15 pg/mL
standard curve range: 8.23-6000 pg/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... PECAM1(18613)

General description

The Human PECAM-1 ELISA (Enzyme-Linked Immunosorbent Assay) kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of human PECAM-1 in serum, plasma, cell culture supernatants and urine.

Immunogen

Recombinant Human PECAM1

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Other Notes

A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABELADAELISA 1X Assay/Sample Diluent Buffer A (Item D1)SDS

  • RABELADBELISA 5X Assay/Sample Diluent Buffer B (Item E1)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andrei A Karpov et al.
International journal of molecular sciences, 22(3) (2021-01-28)
Chronic thromboembolic pulmonary hypertension (CTEPH) is a rare and life-threatening complication of pulmonary embolism. As existing animal models of CTEPH do not fully recapitulate complex disease pathophysiology, we report a new rat model for CTEPH evoked by repetitive embolization of
Zhen Yan et al.
International journal of nanomedicine, 12, 4433-4442 (2017-06-28)
Engineered zinc oxide nanoparticles (ZnO-NPs) are currently being produced in high tonnage. Exposure to ZnO-NPs presents potential risks to cardiovascular system. Thus far, the toxicological effects of ZnO-NPs on cardiovascular system have not been well characterized. In this study, human
Joerg Kellermair et al.
Journal of thrombosis and thrombolysis, 50(2), 380-385 (2020-03-28)
D-Dimer has a high sensitivity but a low specificity for the diagnosis of deep vein thrombosis (DVT) which limits its implementation as a general screening parameter. There is a demand for additional biomarkers to improve its diagnostic accuracy. Soluble platelet
Huiwen Jiang et al.
Annals of hematology, 98(7), 1721-1732 (2019-05-06)
The CD19-targeted chimeric antigen receptor T cell (CAR-T) therapy has been widely proved effective on relapsed and refractory (r/r) B cell acute lymphoblastic leukemia (B-ALL). Meanwhile, CAR-T therapy-related toxicities, including cytokine release syndrome (CRS) and neurological toxicities, are drawing researchers'
Claudia Strobel et al.
Beilstein journal of nanotechnology, 5, 1795-1807 (2014-11-11)
Cerium dioxide (CeO2) and silicon dioxide (SiO2) nanoparticles are of widespread use in modern life. This means that human beings are markedly exposed to them in their everyday life. Once passing biological barriers, these nanoparticles are expected to interact with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service