RAB0157
Cell-Based ELISA for detecting phospho-EGFR (pTyr992) in cultured cell lines
96 assays
About This Item
Recommended Products
usage
96 assays
species reactivity
rat, human, mouse
technique(s)
ELISA: suitable
capture ELISA: suitable
input
sample type cultured cells (96 assays)
shipped in
wet ice
storage temp.
−20°C
Gene Information
human ... EGFR(1965)
General description
Immunogen
Application
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)
Kit Components Also Available Separately
- RABBLOCK5x Blocking Solution (Item F)SDS
- RABFIX1Fixing Solution (Item D)SDS
- RABHRP1HRP-conjugated Anti-Rabbit IgG Concentrate (Item I1)SDS
- RABHRP2HRP-conjugated Anti-Mouse IgG Concentrate (Item I2)SDS
- RABPLATE1Uncoated MicroplateSDS
- RABQUENCHQuenching Solution for Cell-based ELISA Assay (Item E)SDS
- RABSTOP1Stop SolutionSDS
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 1B - Eye Dam. 1 - Met. Corr. 1 - Muta. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service