Key Documents
RA438-EP
Rotor for Eppendorf® 5702/5702R/5702RH Centrifuges
A-4-38, aerosol-tight, without buckets, for round-bottom tubes only
About This Item
UNSPSC Code:
41103909
Pricing and availability is not currently available.
feature
to hold 4 x 100 mL round buckets
to hold 4 x 90 mL rectangular buckets
aerosol-tight
for round-bottom tubes only
swing bucket rotor
without buckets
packaging
pack of 1 ea
manufacturer/tradename
Eppendorf® 022639064 (NA)
Eppendorf® 5702740004 (EU)
parameter
2,750 × g max. RCF
technique(s)
centrifugation: suitable
compatibility
for use with centrifuge models 5702/5702R/502RH
Related Categories
General description
Rotors to fit Eppendorf centrifuge models 5702, 5720 R, and 5720 RH
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)