Skip to Content
Merck
All Photos(1)

Key Documents

R8501

Sigma-Aldrich

Ribonucleic acid-core from baker′s yeast (S. cerevisiae)

Type XI-C

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
41106305

type

Type XI-C

storage temp.

−20°C

Application

Ribonucleic acid (RNA) from baker′s yeast S. cerevisiae may be used as a substrate for studying ribonuclease activities of enzymes such as RNAase(s).

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T V Iamkovaia et al.
Prikladnaia biokhimiia i mikrobiologiia, 42(1), 93-97 (2006-03-09)
A simple method of production of total RNA from baker's yeast was developed. Total RNA was isolated from yeast (Saccharomyces cerevisiae) biomass using lysis with sodium dodecyl sulfate at 100 degrees C for 40-60 min and subsequent precipitation of the
Xiangli Wu et al.
The Journal of general and applied microbiology, 56(3), 231-239 (2010-07-22)
A 15-kDa RNase was purified from Pleurotus djamor using ion exchange chromatography and gel filtration. Its N-terminal amino acid sequence was different from previously reported RNase sequences of mushrooms belonging to the genus of Pleurotus and other genera. The RNase
The preparation and characterization of ribonucleic acids from yeast.
A M CRESTFIELD et al.
The Journal of biological chemistry, 216(1), 185-193 (1955-09-01)
RNA immunoprecipitation for determining RNA-protein associations in vivo.
Gilbert C, Svejstrup JQ.
Current Protocols in Molecular Biology, Unit 27-Unit 27 (2006)
Evandro Fei Fang et al.
FEBS letters, 584(18), 4089-4096 (2010-09-08)
A pumpkin 2S albumin with ribonuclease (RNase) activity was purified from pumpkin seeds (Cucurbita sp.) by liquid chromatographic techniques. It manifested potent RNase activity toward baker's yeast RNA and calf liver RNA, and some polyhomoribonucleotides, including poly(A), poly(U) and poly(C)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service