Skip to Content
Merck
All Photos(1)

Key Documents

R6153

Sigma-Aldrich

Monoclonal Anti-RhoE antibody produced in mouse

~2 mg/mL, clone 4, purified immunoglobulin, aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

4, monoclonal

form

aqueous solution

mol wt

antigen ~29 kDa

species reactivity

human, mouse

concentration

~2 mg/mL

technique(s)

immunocytochemistry: suitable
microarray: suitable
western blot: 4-8 μg/mL using HMVEC total cell extract.

isotype

IgG1

UniProt accession no.

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RND3(390)
mouse ... Rnd3(74194)

Specificity

The antibody weakly cross reacts with Rnd1.

Immunogen

purified recombinant full length RhoE.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Raquel Talens-Visconti et al.
Journal of neurochemistry, 112(4), 1074-1087 (2009-12-09)
Neurite formation involves coordinated changes between the actin cytoskeleton and the microtubule network. Rho GTPases are clearly implicated in several aspects of neuronal development and function. Indeed, RhoA is a negative regulator of neurite outgrowth and its effector Rho-kinase mediates

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service