Skip to Content
Merck
All Photos(1)

Documents

R3026

Sigma-Aldrich

Anti-Rabphilin 3A antibody produced in rabbit

~1 mg/mL, affinity isolated antibody, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~75 kDa

species reactivity

mouse, rat

should not react with

human

concentration

~1 mg/mL

technique(s)

western blot: 1 μg/mL using rat brain extract

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

mouse ... Rph3a(19894)
rat ... Rph3a(171039)

Immunogen

synthetic peptide corresponding to amino acids 1-18 of rat rabphilin 3A. This sequence is completely conserved between rat and mouse rabphilin 3A.

Physical form

Solution in phosphate buffered saline with 1.0 mg/mL bovine serum albumin and 0.05% sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Erwin R Arias-Hervert et al.
Physiological reports, 8(9), e14428-e14428 (2020-05-03)
Members of the Rab3 gene family are considered central to membrane trafficking of synaptic vesicles at mammalian central excitatory synapses. Recent evidence, however, indicates that the Rab27B-GTPase, which is highly homologous to the Rab3 family, is also enriched on SV

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service