Skip to Content
Merck
All Photos(1)

Key Documents

R2152

Sigma-Aldrich

Rap1A human

buffered aqueous glycerol solution, recombinant, expressed in E. coli, GST-tagged

Synonym(s):

Ras Related Protein 1A

Sign Into View Organizational & Contract Pricing

Select a Size

25 μG
R$3,928.00

R$3,928.00


Estimated to ship onMay 15, 2025



Select a Size

Change View
25 μG
R$3,928.00

About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.32

R$3,928.00


Estimated to ship onMay 15, 2025


recombinant

expressed in E. coli

Quality Level

form

buffered aqueous glycerol solution

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... RAP1A(5906)

Application

Can be used as a control in immunoblotting and in protein binding assays.

Biochem/physiol Actions

A small Ras-like GTP-binding protein (21 kDa, native).
Rap1A, a small Ras-like GTP-binding protein (21 kDa, native), is a potential modulator of the Ras/Raf/MAP kinase intracellular signaling cascade.

Other Notes

GST-tagged Rap1A fusion protein

Physical form

Supplied in 50 mM Tris, pH 7.5, containing 150 mM sodium chloride, 10 mM magnesium chloride, 1 mM DTT, and 10% glycerol.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Y Takai et al.
Physiological reviews, 81(1), 153-208 (2001-01-12)
Small GTP-binding proteins (G proteins) exist in eukaryotes from yeast to human and constitute a superfamily consisting of more than 100 members. This superfamily is structurally classified into at least five families: the Ras, Rho, Rab, Sar1/Arf, and Ran families.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service