Skip to Content
Merck
All Photos(1)

Key Documents

R123

Sigma-Aldrich

RBI 257 maleate salt

solid

Synonym(s):

1-[4-Iodobenzyl]-4-[[2-[3-isopropoxy]pyridyl]-methylamino]piperidine maleate salt, p-Iodo-U-101,958 maleate salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H28IN3O · C4H4O4
Molecular Weight:
581.44
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

form

solid

color

white

solubility

H2O: 0.2 mg/mL
ethanol: 1 mg/mL
0.1 M HCl: 7 mg/mL
DMSO: 9 mg/mL

storage temp.

2-8°C

SMILES string

[H]\C(=C(/[H])C(O)=O)C(O)=O.CC(C)Oc1cccnc1N(C)C2CCN(CC2)Cc3ccc(I)cc3

Biochem/physiol Actions

High affinity D4 dopamine receptor ligand.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N S Kula et al.
European journal of pharmacology, 331(2-3), 333-336 (1997-07-23)
RBI-257 (1-[4-iodobenzyl]-4-[N-(3-isopropoxy-2-pyridinyl)-N-methyl]-aminopiperid ine), the p-iodobenzyl analog of U-101,958 (1-benzyl-4-[N-(3-isopropoxy-2-pyridinyl)-N-methyl]-aminopiperidine) had a lower dissociation constant (Ki = 0.3 vs. 2.7 nM) and higher selectivity than U-101,958 at dopamine D4 receptors, over dopamine D2 and D3 receptors in transfected cell membranes and
Compounds selective for dopamine receptor subtypes.
Kebabian, et al.
Drug Discovery Today, 2, 333-340 (1997)
F Henry et al.
Theriogenology, 83(8), 1373-1380 (2015-03-05)
From many endangered or threatened species which are expected to profit from assisted reproduction techniques, mainly epididymal sperm of dead or freshly castrated males are available. These sperm had contact to epididymal secretion products but not to seminal fluid components.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service