Q0259
Q-Ceramic HyperD® F
50 μm mean particle size
Synonym(s):
HyperD® ceramic ion exchangers
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
mean particle size
50 μm
capacity
>0.20 meq/mL
storage temp.
2-8°C
Looking for similar products? Visit Product Comparison Guide
Application
Ceramic HyperD media are used in protein chromatography and affinity chromatography. Ceramic HyperD media have been used in the development of a capture step of a human recombinant F(ab′)(2) produced and expressed in baculovirus-infected cells and to identify whey proteins as a model system for chromatographic separation of proteins.
Features and Benefits
HyperD media are highly porous, rigid ceramic beads filled with a functionalized hydrophilic gel. The rigid bead allows extremely high linear flow rates without bed compression. The hydrogel exchanges throughout its volume, not just on the surface. And the media do not change volume due to changes in pH or ionic strength.
The supports are stable in commonly used solvents and alkaline or acid environments. Supplied as aqueous suspensions in 1 M NaCl with 20% ethanol to inhibit bacterial growth.
The supports are stable in commonly used solvents and alkaline or acid environments. Supplied as aqueous suspensions in 1 M NaCl with 20% ethanol to inhibit bacterial growth.
Legal Information
HyperD is a registered trademark of Pall Corporation
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1 - Flam. Liq. 3 - Skin Irrit. 2
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
95.0 °F
Flash Point(C)
35 °C
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Journal of clinical oncology : official journal of the American Society of Clinical Oncology, 27(17), 2787-2792 (2009-05-06)
There are no reliable blood biomarkers to detect early lung cancer. We used a novel strategy that allows discovery of differentially present proteins against a complex and variable background. Mass spectrometry analyses of paired pulmonary venous-radial arterial blood from 16
Molecular medicine (Cambridge, Mass.), 14(7-8), 383-394 (2008-05-23)
Hypertension represents one of the main risk factors for vascular diseases. Genetic susceptibility may influence the rate of its development and the associated vascular remodeling. To explore markers of hypertension-related morbidity, we have used surface-enhanced laser desorption/ionization time-of-flight (SELDI-TOF) mass
Neurobiology of disease, 23(1), 61-76 (2006-03-22)
This paper describes the high-throughput proteomic analysis of the dorsolateral prefrontal cortex (DLPFC) from schizophrenia (SCHIZ), bipolar (BD), and normal control cohorts from the Harvard Brain Tissue Resource Center performed using ProteinChip technology based on the surface-enhanced laser desorption/ionization time
Methods in molecular biology (Clifton, N.J.), 357, 331-341 (2006-12-19)
Surface-enhanced laser desorption/ionization time-of-flight mass spectrometry (TOFMS) allows for rapid differential proteomic analysis of various experimental conditions. In the first step of retention chromatography, the proteome to be analyzed is fractionated based on the biochemical properties of the proteins or
Blood, 111(3), 1201-1207 (2007-10-05)
Early tumor detection and intervention are important determinants of survival in patients with cancer. We have recently reported that the "platelet angiogenesis proteome" may be used to detect microscopic tumors in mice. We now present evidence that changes in platelet-associated
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service