Skip to Content
Merck
All Photos(1)

Key Documents

PZ0403

Sigma-Aldrich

PF-9366

≥98% (HPLC)

Synonym(s):

7-Chloro-N,N-dimethyl-5-phenyl-[1,2,4]triazolo[4,3-a]quinoline-1-ethanamine, PF 9366, PF9366

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
R$700.00
25 MG
R$2,821.00

R$700.00


Available to ship onApril 07, 2025Details


Request a Bulk Order

Select a Size

Change View
5 MG
R$700.00
25 MG
R$2,821.00

About This Item

Empirical Formula (Hill Notation):
C20H19ClN4
CAS Number:
Molecular Weight:
350.84
MDL number:
UNSPSC Code:
51111800
NACRES:
NA.77

R$700.00


Available to ship onApril 07, 2025Details


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

room temp

SMILES string

ClC(C=C1)=CC(C(C2=CC=CC=C2)=C3)=C1N4C3=NN=C4CCN(C)C

InChI

1S/C20H19ClN4/c1-24(2)11-10-19-22-23-20-13-16(14-6-4-3-5-7-14)17-12-15(21)8-9-18(17)25(19)20/h3-9,12-13H,10-11H2,1-2H3

InChI key

LYLASWLQCMKZAT-UHFFFAOYSA-N

Biochem/physiol Actions

PF-9366 is a methionine adenosyltransferase 2A allosteric inhibitor (human Mat2A IC50/Kd = 420 nM/170 nM) whose binding site overlaps with that of the Mat2A regulator, Mat2B. PF-9366 inhibits cellular SAM production in a Mat2B-competitive manner (IC50 post 6-hr treatment = 1.2 μM wihout vs. 0.86 μM with Mat2B knockdown; H520 lung carcinoma cells) without antiproliferation efficacy in cancer cultures due to an induction of Mat2A upregulation upon allosteric inhibition. Similar to Mat2B, PF-9366 allosteric binding alters Mat2A active site, causing increased substrate affinity and decreased enzyme turnover.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Weiwei Yu et al.
Molecular cell, 75(6), 1147-1160 (2019-08-20)
Activated macrophages adapt their metabolic pathways to drive the pro-inflammatory phenotype, but little is known about the biochemical underpinnings of this process. Here, we find that lipopolysaccharide (LPS) activates the pentose phosphate pathway, the serine synthesis pathway, and one-carbon metabolism
Jiraporn Panmanee et al.
The FEBS journal, 286(11), 2135-2154 (2019-02-19)
Methylation is an underpinning process of life and provides control for biological processes such as DNA synthesis, cell growth, and apoptosis. Methionine adenosyltransferases (MAT) produce the cellular methyl donor, S-Adenosylmethionine (SAMe). Dysregulation of SAMe level is a relevant event in
Casey L Quinlan et al.
Nature chemical biology, 13(7), 785-792 (2017-05-30)
S-Adenosyl-L-methionine (SAM) is an enzyme cofactor used in methyl transfer reactions and polyamine biosynthesis. The biosynthesis of SAM from ATP and L-methionine is performed by the methionine adenosyltransferase enzyme family (Mat; EC 2.5.1.6). Human methionine adenosyltransferase 2A (Mat2A), the extrahepatic

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service