Skip to Content
Merck
All Photos(1)

Key Documents

PZ0332

Sigma-Aldrich

ISOX-INACT

≥98% (HPLC)

Synonym(s):

4-(2-(5-(3,5-Dimethylisoxazol-4-yl)-2-(4-methoxyphenethyl)-4,6-dimethyl-1Hbenzo[d]imidazol-1-yl)ethyl)morpholine

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
R$2,303.00

R$2,303.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
R$2,303.00

About This Item

Empirical Formula (Hill Notation):
C29H36N4O3
CAS Number:
Molecular Weight:
488.62
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

R$2,303.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 5 mg/mL, clear (warmed)

storage temp.

room temp

SMILES string

CC1=NOC(C)=C1C2=C(C)C(N=C(CCC3=CC=C(OC)C=C3)N4CCN5CCOCC5)=C4C=C2C

InChI

1S/C29H36N4O3/c1-19-18-25-29(20(2)27(19)28-21(3)31-36-22(28)4)30-26(11-8-23-6-9-24(34-5)10-7-23)33(25)13-12-32-14-16-35-17-15-32/h6-7,9-10,18H,8,11-17H2,1-5H3

InChI key

FTOHXUPHICJSCR-UHFFFAOYSA-N

Biochem/physiol Actions

CBP BRD (CREB binding protein bromodomain) inhibition serves as an effective therapeutic target for Parkinson′s disease. The inhibition is followed by negative regulation of a number of macrophage inflammatory genes. It also downregulates G-protein signaling 4 mRNA.[1]
ISOX-INACT is an inactive control probe for the highly-selective inhibitor of the bromodomain of CREB binding protein (CBP BRD) PF-CBP1.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eugene L Piatnitski Chekler et al.
Chemistry & biology, 22(12), 1588-1596 (2015-12-17)
Bromodomains are involved in transcriptional regulation through the recognition of acetyl lysine modifications on diverse proteins. Selective pharmacological modulators of bromodomains are lacking, although the largely hydrophobic nature of the pocket makes these modules attractive targets for small-molecule inhibitors. This

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service