Skip to Content
Merck
All Photos(1)

Key Documents

PZ0309

Sigma-Aldrich

PF-04634817 succinate

≥98% (HPLC)

Synonym(s):

1,5-Anhydro-2,3-dideoxy-4-O-methyl-3-[[(1R,3S)-3-(1-methylethyl)-3-[[(1S,4S)-5-[6-(trifluoromethyl)-4-pyrimidinyl]-2,5-diazabicyclo[2.2.1]hept-2-yl]carbonyl]cyclopentyl]amino]-D-erythro-pentitol succinate, PF 04634817 succinate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H36F3N5O3 · C4H6O4
CAS Number:
Molecular Weight:
629.67
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

H2O: 30 mg/mL, clear

storage temp.

room temp

SMILES string

CC(C)[C@@]1(C(N2C[C@@]3([H])N(C4=NC=NC(C(F)(F)F)=C4)C[C@]2([H])C3)=O)C[C@H](N[C@H]5CCOC[C@H]5OC)CC1.O=C(CCC(O)=O)O

InChI

1S/C25H36F3N5O3.C4H6O4/c1-15(2)24(6-4-16(10-24)31-19-5-7-36-13-20(19)35-3)23(34)33-12-17-8-18(33)11-32(17)22-9-21(25(26,27)28)29-14-30-22;5-3(6)1-2-4(7)8/h9,14-20,31H,4-8,10-13H2,1-3H3;1-2H2,(H,5,6)(H,7,8)/t16-,17+,18+,19+,20-,24+;/m1./s1

InChI key

WRPPUEHVLSHXJG-HTQYHTRRSA-N

Biochem/physiol Actions

PF-04634817 is a potent and selective antagonist of CC chemokine receptor 2 and 5 (CCR2/5).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service