Skip to Content
Merck
All Photos(1)

Key Documents

PZ0228

Sigma-Aldrich

PF-04628935

≥98% (HPLC)

Synonym(s):

1-[2-[[2-Chloro-4-(2H-1,2,3-triazol-2-yl)phenyl]methyl]-2,7-diazaspiro[3.5]non-7-yl]-2-(2-methylimidazo[2,1-b]thiazol-6-yl)-ethanone, PF-4628935

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
R$670.00
25 MG
R$2,614.00

R$670.00


Estimated to ship onMay 23, 2025


Request a Bulk Order

Select a Size

Change View
5 MG
R$670.00
25 MG
R$2,614.00

About This Item

Empirical Formula (Hill Notation):
C24H26ClN7OS
CAS Number:
Molecular Weight:
496.03
UNSPSC Code:
51111800
NACRES:
NA.77

R$670.00


Estimated to ship onMay 23, 2025


Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 10 mg/mL, clear

storage temp.

room temp

Biochem/physiol Actions

PF-04628935 is a potent antagonist/inverse agonist of the ghrelin receptor, growth hormone secretagogue receptor 1a (GHS-R1a) with an IC50 of 4.6 nM. Ghrelin and GSHR are involved in regulation of food intake and long-term energy homeostasis; GSHR ligands are of interest for obesity and other metabolic disorders. PF-04628935 is orally bioavailable and brain penetrant.
PF-04628935 is a potent antagonist/inverse agonist of the ghrelin receptor, growth hormone secretagogue receptor 1a (GHS-R1a).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service