Skip to Content
Merck
All Photos(1)

Key Documents

PZ0156

Sigma-Aldrich

UK-383,367

≥98% (HPLC)

Synonym(s):

(βR)-3-(Aminocarbonyl)-β-(3-cyclohexylpropyl)-N-hydroxy-1,2,4-oxadiazole-5-propanamide, 5-{(1R)-4-Cyclohexyl-1-[2-(hydroxyamino)-2-oxoethyl]butyl}-1,2,4-oxadiazole-3-carboxamide

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
R$1,235.00
25 MG
R$4,693.00

R$1,235.00


Estimated to ship onJune 02, 2025


Request a Bulk Order

Select a Size

Change View
5 MG
R$1,235.00
25 MG
R$4,693.00

About This Item

Empirical Formula (Hill Notation):
C15H24N4O4
CAS Number:
Molecular Weight:
324.38
MDL number:
UNSPSC Code:
41121800
PubChem Substance ID:
NACRES:
NA.77

R$1,235.00


Estimated to ship onJune 02, 2025


Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

optical activity

[α]/D 14.0 to 23.0°, c = 0.5 in methanol

color

white to off-white

solubility

DMSO: >30 mg/mL

storage temp.

room temp

SMILES string

NC(=O)c1noc(n1)[C@H](CCCC2CCCCC2)CC(=O)NO

InChI

1S/C15H24N4O4/c16-13(21)14-17-15(23-19-14)11(9-12(20)18-22)8-4-7-10-5-2-1-3-6-10/h10-11,22H,1-9H2,(H2,16,21)(H,18,20)/t11-/m1/s1

InChI key

ARJCBSRIPGJMAD-LLVKDONJSA-N

Biochem/physiol Actions

Procollagen C-proteinase (PCP) inhibitor.
UK-383,367 is a procollagen C-proteinase (PCP) inhibitor.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yael Eshed-Eisenbach et al.
Neuron, 106(5), 806-815 (2020-03-27)
During development of the peripheral nervous system (PNS), Schwann-cell-secreted gliomedin induces the clustering of Na+ channels at the edges of each myelin segment to form nodes of Ranvier. Here we show that bone morphogenetic protein-1 (BMP1)/Tolloid (TLD)-like proteinases confine Na+

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service